Leak diverters for pipes, PIG®
Supplier: NEW PIG
Certificates
About this item
Purpose-built design catches pipe leaks or condensation to protect equipment, inventory and personnel.
- Hangs from a leaking pipe to catch nuisance leaks and channel them to a floor drain or collection container
- Non-permeable tarpaulin is vinyl coated for strength and durability
- Resists punctures, UV and mildew for long life
- Built-in grommets and included rubber straps make it easy to hang from an overhead pipe
Flow rate of 12,5 L per minute when used with a ½" ID hose.