Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poly(2-ethyl-2-oxazoline), M.W. ~200,000
Poly(2-ethyl-2-oxazoline), M.W. ~50,000, Sigma-Aldrich®
Poly(2-ethyl-2-oxazoline), M.W. ~50,000, Sigma-Aldrich®
Poly(2-ethyl-2-oxazoline), M.W. ~50,000, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Poly(2-ethyl-2-oxazoline), M.W. ~50,000, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
25805-17-8
:
161358-47-0
Formula:
(C₅H₉NO)x
:
200000 g/mol
:
M-Clarity™ quality level = MQ200