6-Bromo-2-naphthoic acid
Catalog # MOLE56202798-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:251.08 g/mol
CAS Number:5773-80-8
Formula:C₁₁H₇BrO₂
MDL Number:MFCD01075720