Order Entry
Germany
ContactUsLinkComponent
L(+)-Glutamine 0.2 mol/l in aqueous solution, Sigma-Aldrich®
L(+)-Glutamine 0.2 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALG2150-20ML
Supplier:  Merck
CAS Number:  
undefined
L(+)-Glutamine 0.2 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALG2150-20ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    20 mL
  • Seq:
    2305

Specifications

About this item

 :
 :146.15 g/mol
Merck Index:13,04484
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:200-292-1
CAS Number:56-85-9
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044