Pesticide reference standard, Pentachlorophenyl acetate 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA15971000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1441-02-7
- Description:Pentachlorophenyl acetate 100 µg/ml in cyclohexane
- Formula:C₈H₃Cl₅O₂
- Description:Pentachlorophenyl acetate