Order Entry
Germany
Orders LinkContactUsLinkComponent
Progesterone

Progesterone

Glass bottle for solids, 100 mg
Catalog # PROOICRS0400
Progesterone
Catalog # PROOICRS0400
CAS Number:  57-83-0
Progesterone

Glass bottle for solids, 100 mg

CAS Number:  57-83-0
Price
235,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:57-83-0
  • Formula:C₂₁H₃₀O₂
  • MDL Number:MFCD00003658
  • EINECS:200-350-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications