Progesterone
1.000
Catalog # FITZ20-PR20
Supplier: Fitzgerald Industries
Jump to:
Product Details & Documents
- CAS Number:57-83-0
- Formula:C₂₁H₃₀O₂
- MDL Number:MFCD00003658
- EINECS:200-350-6
Specifications
Specifications
- Pk:1.000
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



