Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Dibenzofuranboronic acid
4-Dibenzofuranboronic acid
4-Dibenzofuranboronic acid
4-Dibenzofuranboronic acid
FLUO011026-5G
:
FLUOROCHEM
:
4-Dibenzofuranboronic acid
FLUO011026-5G
:
FLUOROCHEM
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
212.01 g/mol
:
100124-06-9
:
C₁₂H₉BO₃
:
MFCD00092336