Order Entry
Germany
ContactUsLinkComponent
Diethyl (2,2-diethoxyethyl)phosphonate, Sigma-Aldrich®
Diethyl (2,2-diethoxyethyl)phosphonate, Sigma-Aldrich®
Catalog # SIALD99250-25G
Supplier:  Merck
CAS Number:  
Diethyl (2,2-diethoxyethyl)phosphonate, Sigma-Aldrich®
Catalog # SIALD99250-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :254.26 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:231-504-0
CAS Number:7598-61-0
Formula:C₁₀H₂₃O₅P
MDL Number:MFCD00009236