
Diethyl (2,2-diethoxyethyl)phosphonate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7598-61-0
:
Formula:C₁₀H₂₃O₅P
MDL Number:MFCD00009236
:254.26 g/mol
Boiling Pt:101…103 °C (8 mmHg)
Density:1.052
Flash Pt:>110 °C (230 °F)
EINECS:231-504-0
:GHS07
Related Information
Frequently Bought Together