Order Entry
Germany
ContactUsLinkComponent
1,2-Naphthoquinone-4-sulphonic acid sodium salt, Sigma-Aldrich®
1,2-Naphthoquinone-4-sulphonic acid sodium salt, Sigma-Aldrich®
Catalog # SIAL226017-10G
Supplier:  Merck
CAS Number:  
1,2-Naphthoquinone-4-sulphonic acid sodium salt, Sigma-Aldrich®
Catalog # SIAL226017-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :260.20 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:208-308-9
CAS Number:521-24-4
Formula:C₁₀H₅NaO₅S
MDL Number:MFCD00001700