Ethyl dichlorophosphite
1
Catalog # CHMPFL13573.POR
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: Ethyl phosphorodichloridite
- CAS Number:1498-42-6
- Formula:C₂H₅Cl₂OP
- MDL Number:MFCD00000525
- Molecular Weight:146.94 g/mol
- EINECS:216-098-5
- UN:2924
- ADR:3,II
Specifications
Specifications
- Pk:1
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



