Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxy-2-methylpropiophenone
2-Hydroxy-2-methylpropiophenone, Sigma-Aldrich®
2-Hydroxy-2-methylpropiophenone, Sigma-Aldrich®
2-Hydroxy-2-methylpropiophenone, Sigma-Aldrich®
Supplier:
Merck
Certificates
Bild vergrößern
2-Hydroxy-2-methylpropiophenone, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
7473-98-5
:
Formula:
C₁₀H₁₂O₂
MDL Number:
MFCD00040710
:
164.2000 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
231-272-0