Order Entry
Germany
ContactUsLinkComponent
Phloretin, Millipore®
Phloretin, Millipore®
Catalog # 524488-200
CAS Number:  
Phloretin, Millipore®
Catalog # 524488-200
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 mg
  • Seq:
    2530

Specifications

About this item

 :
 :274.28 g/mol
EINECS:200-488-7
CAS Number:60-82-2
Formula:C₁₅H₁₄O₅
MDL Number:MFCD00002288