(R)-(-)-Nipecotic acid
Catalog # MOLE22454408-100MG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:129.16 g/mol
CAS Number:25137-00-2
Formula:C₆H₁₁NO₂
MDL Number:MFCD01630787