Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, Benzo[e]pyrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, Benzo[e]pyrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20645000CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Benzo[e]pyrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20645000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    192-97-2
  • Description:
    Benzo[e]pyrene 10 µg/ml in cyclohexane
  • Formula:
    C₂₀H₁₂
  • Description:
    Benzo[e]pyrene

Specifications

About this item