Order Entry
Germany
ContactUsLinkComponent
Diethylene glycol methyl ethyl ether for synthesis, Sigma-Aldrich®
Diethylene glycol methyl ethyl ether for synthesis, Sigma-Aldrich®
Catalog # 8.43827.1000
Supplier:  Merck
CAS Number:  
Diethylene glycol methyl ethyl ether for synthesis, Sigma-Aldrich®
Catalog # 8.43827.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    76 g/l (25 °C)
  • Melting Point:
    -72 °C
  • Molar Mass:
    148.2 g/mol
  • Flash Point:
    82 °C
  • Density:
    0.925 g/cm3

Specification Test Results

About this item

 :
 :148.2001 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:213-690-5
CAS Number:1002-67-1
Formula:C₇H₁₆O₃
MDL Number:MFCD01727263