Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Ribonuclease A
Ribonuclease A
Ribonuclease A
Supplier:
OMEGA BIO-TEK
Certificates
Expand image
Ribonuclease A
Supplier:
OMEGA BIO-TEK
Certificates
About this item
Enzyme activity is 100 mg/ml (for 5 ml)
Specifications
CAS No.:
9001-99-4
Storage conditions:
Freezer
MDL No.:
MFCD00132184
Enzyme name:
Ribonuclease A
Enzyme synonyms:
RNase A
Specifications
Related Information
Documentation
Certificates
Search by Lot Number
SDS
sds_VWRCPD090
sds_VWRCRNA-03
Documentation