0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Enzymes
Ribonuclease A
Ribonuclease A
Catalog # AMBI2274
Supplier:
Ambion
CAS Number:
Expand image
Ribonuclease A
Catalog # AMBI2274
Supplier:
Ambion
CAS Number:
Specifications
Pk:
200 µl
CAS No.:
9001-99-4
Storage conditions:
Freezer
MDL No.:
MFCD00132184
Enzyme name:
Ribonuclease A
Enzyme synonyms:
RNase A
Specifications
About this item
Product Details
Product Details