Thiazolidine-2-carboxylic acid
Catalog # MAYBKM05539.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:133.169 g/mol
CAS Number:65126-70-7
:16310-13-7
Formula:C₄H₇NO₂S
MDL Number:MFCD00068005