Sodium dihydrogen phosphate
Glass bottle for solids, 1 kg
Catalog # RC-098
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
- CAS Number:7558-80-7
- Formula:NaH₂PO₄
- MDL Number:MFCD00003527
- Molecular Weight:119.98 g/mol
- Merck Index:12,08806
- Melting Pt:∼650 °C
- Density:0.9996 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-449-2
- REACH:01-2119489796-13
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



