

Lithium perchlorate ≥98%
Catalog # A16059.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:106.39 g/mol
Merck Index:13,05561
Boiling Pt:430 °C (1013 hPa)
Melting Pt:236…237 °C
Density:2.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-237-2
UN:1481
ADR:5.1,II
:GHS03
:GHS07
CAS Number:7791-03-9
Formula:ClLiO₄
MDL Number:MFCD00011079