Calcium chloride dihydrate
RC-031
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
:
:147.01 g/mol
:13,01660
:1600 °C (1013 hPa)
:176 °C
:1.85 g/cm³ (20 °C)
:Ambient
:233-140-8
:GHS07
:10035-04-8
:CaCl₂·2H₂O
:MFCD00149613