Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Phenyl-1,3-propanediol
2-Phenyl-1,3-propanediol ≥98%
2-Phenyl-1,3-propanediol ≥98%
:
Thermo Fisher Scientific
2-Phenyl-1,3-propanediol ≥98%
:
Thermo Fisher Scientific
:
1570-95-2
:
:
HOH₂CCH(C₆H₅)CH₂OH
:
MFCD00236056
:
152.19 g/mol
:
53…56 °C
:
411-810-2
:
GHS05
:
Ambient
msds_ALFAH56761
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years