
4-Bromopyridinium chloride
Catalog # TOROB686735-100G
Supplier: TORONTO RESEARCH CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:194.46 g/mol
Melting Pt:235…238 °C
EINECS:243-128-4
UN:2811
ADR:6.1,III
:GHS07
CAS Number:19524-06-2
Formula:C₅H₅BrClN
MDL Number:MFCD00012828