1-Hexyl-3-methylimidazolium bromide
Catalog # UPTICH577
Supplier: Uptima
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:247.18 g/mol
CAS Number:85100-78-3
Formula:C₁₀H₁₉BrN₂
MDL Number:MFCD03427612