Platinum ≥99.98% (metals basis), powder -200/+400 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-06-4
:
Formula:Pt
MDL Number:MFCD00011179
:195.08 g/mol
Boiling Pt:4530 °C (1013 hPa)
Melting Pt:1773 °C
Density:21.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-116-1
UN:3089
ADR:4.1,II