
Guanidinium thiocyanate
Supplier: G-Biosciences
Certificates
About this item
CAS Number:593-84-0
:
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027
:118.16 g/mol
Melting Pt:118 °C
Density:1.29 g/cm³ (20 °C)
EINECS:209-812-1
:GHS07
Storage Temperature:Ambient