Order Entry
Germany
Orders LinkContactUsLinkComponent
Pipette controller, FastPette™ V2

Pipette controller, FastPette™ V2

Electronic pipette controllers, for glass or plastic pipettes, EU-plug, 1 - 100 ml
Catalog # LBNEP2000-230EU
Pipette controller, FastPette™ V2
Catalog # LBNEP2000-230EU
Pipette controller, FastPette™ V2

Electronic pipette controllers, for glass or plastic pipettes, EU-plug, 1 - 100 ml

Price
Inquire For Price
Log in for availability
1 * 1 Pack.
Product Details & Documents

This battery powered pipette controller works with glass and plastic pipettes in the 1 to 100 ml range.

  • Lightweight handle with smooth push-buttons and ergonomically located switches
  • High aspiration speed - 25 ml in 4 seconds
  • Cordless work for up to 8 continuous hours
  • Aspiration speed can be set to high or low to adjust for the volume of the pipette and viscosity of the liquid
  • Suction can be adjusted by the amount of pressure applied to the trigger button
  • Dispensing can be carried out by gravity or supported by the pump, which empties the pipette with a blow out step
  • Protected from the entrance of liquids by both a PTFE filter and a safety valve
  • Filter and pipette holder are easily exchanged and autoclaved

Delivery information: Supplied with power supply/charger and PTFE filter.

Specifications
  • Capacity:
    1 - 100 ml
  • Description:
    Electronic pipette controllers, for glass or plastic pipettes, EU-plug
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.