Concentrator
Supplier: G-Biosciences
Certificates
About this item
The concentrator is a high molecular weight polymer which will not migrate across the dialysis membrane. The concentrator is placed against the dialysis membrane of the Tube-O-DIALYZER™, where it rapidly absorbs water from the sample and reduces the sample volume. The concentrator can be used with dialysis bags and other dialysis devices.