Orders LinkContactUsLinkComponent
Measuring scoops
 :   VITLAB
Measuring scoops
 :  VITLAB
Measuring scoops
Measuring scoops
PP, coloured. With precision formed filling edge and comfortable, stable handle.
  • Also suitable as weighing scoops
  • Easily readable volume quantities visible on the upper side of the handle
  • Food-safe product according to EC Directive No. 10/2011
Product Family Options
SET
  • Description
    Capacity
    Colour
  • Set of 8 coloured scoops (white, red, grey, black, yellow, dark blue, light blue, green)
    100 ml
    Assorted
    1 * 1 ST
    1 * 1 ST - 15,40 €
    15,40 €
    1 * 1 ST
       15,40 €
  • Description
    Capacity
    Colour
  • 100 ml
    Yellow
    1 * 12 ST
    1 * 12 ST - 20,60 €
    20,60 €
    1 * 12 ST
       20,60 €
  • 100 ml
    Red
    1 * 12 ST
    1 * 12 ST - 20,60 €
    20,60 €
    1 * 12 ST
       20,60 €
  • 100 ml
    Grey
    1 * 12 ST
    1 * 12 ST - 20,60 €
    20,60 €
    1 * 12 ST
       20,60 €
  • 100 ml
    Black
    1 * 12 ST
    1 * 12 ST - 20,60 €
    20,60 €
    1 * 12 ST
       20,60 €
  • 100 ml
    Dark blue
    1 * 12 ST
    1 * 12 ST - 20,60 €
    20,60 €
    1 * 12 ST
       20,60 €
  • 100 ml
    Green
    1 * 12 ST
    1 * 12 ST - 20,60 €
    20,60 €
    1 * 12 ST
       20,60 €
  • 100 ml
    Light blue
    1 * 12 ST
    1 * 12 ST - 20,60 €
    20,60 €
    1 * 12 ST
       20,60 €