Chemical resistant overalls, Mutex Light+
Supplier: HONEYWELL SAFETY
Certificates
About this item
These disposable overalls made of PP laminated with a microporous PE film are designed to protect against particles contamination and chemical splashes.
- Elasticated 3-piece hood, waist, wrists and ankles
- Special crotch design
- Outside flap and dual slider zipper
- Silicone and latex-free
- Anti-static and radioactive particles certified
Typical applications include: Mechanical works, maintenance on site, industrial cleaning in wet environments.
Weight: 65 g/m².
Certifications: Type 5 + 6; Cat. III
Specifications
- Catalog No:
- DALL4500501/S
- DALL4500501/XXXL
- DALL4500501/XL
- DALL4500501/XXL
- DALL4500501/M
- Model:
- Mutex Light
- Mutex Light
- Mutex Light
- Mutex Light
- Mutex Light
- Anti-static:
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Weight:
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²