ContactUsLinkComponent
Centrifuge tubes, conical, without rim
  HECH40939002
undefined
Centrifuge tubes, conical, without rim
  HECH40939002
  • Closure type:
    Without cap
  • Ø×L:
    17×112 mm
  • Capacity:
    15 ml
  • Pk:
    100
AR®-Glass (soda glass) or DURAN®, borosilicate glass.
  • Ffused white graduation
  • Good chemical resistance
Conical bottom, rimless.