Cleanroom anti-fatigue mats, Ergomat® Hygiene
: ERGOMAT
Ergomat® Hygiene is made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, quality issues and sick leave. It is ideal for areas where heat, oil and petrochemical resistance is crucial.
- Upper side with cones
- Can be autoclaved (610×915 mm only) at 100 °C for up to 5 minutes
- Frequent washes or chemical exposure will not affect the state of the mat; no curling up, swelling or expanding
- Moulded, bevelled edges prevent curling, eliminating trip hazards
- Raw, virgin material guarantee durability and ergonomic benefits
- Free from silicone and latex
Suitable for use in Class ISO 5 (100) cleanrooms.
Colour: Blue, green, grey or red
Meets EN ISO/USDA/CFIA standards
: EN ISO/USDA/CFIA