Microcentrifuge tubes
KIMA18166
- Type:with attached snap cap, Eppendorf type
- Version:graduated, conical bottom, non sterile
- Capacity:1,5 ml
- Pk:1.000
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
- Non sterile
With attached push-on cap.