ContactUsLinkComponent
Blood collection tubes with KF+Na2EDTA
Blood collection tubes with KF+Na2EDTA
Catalog # KIMA18506
Blood collection tubes with KF+Na2EDTA
Catalog # KIMA18506
Specifications
  • Ø:
    12 mm
  • Description:
    Blood Collection tube
  • Length:
    86 mm
  • Material:
    PP
  • Cap colour:
    Orange
  • Pk:
    10.000
  • Additive:
    KF+Na2EDTA
Specifications
About this item
PP, translucent.
  • KF+Na2 EDTA for 2,5ml of blood
  • Non sterile
  • With writing area