Order Entry
Germany
Orders LinkContactUsLinkComponent
 

O-Methylhydroxylamine hydrochloride

Supplier:   Cayman Chemical
O-Methylhydroxylamine hydrochloride
Supplier:  Cayman Chemical
Certificates
O-Methylhydroxylamine hydrochloride

2 Options Available Below

Product Details & Documents
Synonyms:  Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride
  • CAS Number:593-56-6
  • Formula:CH₆ClNO
  • MDL Number:MFCD00012951
  • Molecular Weight:83.515 g/mol
  • EINECS:209-798-7
  • UN:3261
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    0,1 g
    Log in for availability
    Log in for availability
    1 * 0,1 g
    1 * 0,1 g - 18,00 €
    18,00 €
    1 * 0,1 g
       18,00 €

  • Glass bottle for solids
    0,5 g
    Log in for availability
    Log in for availability
    1 * 0,5 g
    1 * 0,5 g - 52,00 €
    52,00 €
    1 * 0,5 g
       52,00 €