4-(Ethylthio)benzhydrol
Catalog # FLUO022428-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:842140-67-4
Formula:C₁₅H₁₆OS
MDL Number:MFCD06201259