Order Entry
Germany
ContactUsLinkComponent
 
Mucobromic acid
Catalog # MOLE30334240-100G
Supplier:  Molekula
CAS Number:  
Mucobromic acid
Catalog # MOLE30334240-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :257.88 g/mol
EINECS:207-670-5
UN:3261
ADR:8,III
CAS Number:488-11-9
Formula:C₄H₂Br₂O₃
MDL Number:MFCD00063745