Mucobromic acid 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:488-11-9
:
Formula:C₄H₂Br₂O₃
MDL Number:MFCD00063745
:257.88 g/mol
Melting Pt:122…125 °C
Storage Temperature:Ambient
EINECS:207-670-5
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:122 to 125 °C
- Titration with NaOH:≥99.0 %