Order Entry
Germany
ContactUsLinkComponent
Diethyl (2-cyanoethyl)malonate
Diethyl (2-cyanoethyl)malonate
Catalog # MOLEM83241955
Supplier:  Molekula
CAS Number:  
Diethyl (2-cyanoethyl)malonate
Catalog # MOLEM83241955
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :213.23 g/mol
EINECS:241-260-7
CAS Number:17216-62-5
Formula:C₁₀H₁₅NO₄
MDL Number:MFCD00001966