Order Entry
Germany
ContactUsLinkComponent
Trichloromethanesulphenyl chloride 97%
Toxic
Corrosive
Trichloromethanesulphenyl chloride 97%
Certificates
undefined
Toxic
Corrosive
Trichloromethanesulphenyl chloride 97%

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5370 to 1.5410 (20°C, 589 nm)
  • Assay:
    ≥96.0 %

Specification Test Results

Related Information