2-Chloroethanesulphonic acid sodium salt hydrate
Catalog # MOLE39693672-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:184.57 g/mol
EINECS:241-935-6
CAS Number:1089719-72-1
:15484-44-3
Formula:C₂H₄ClNaO₃S.H₂O
MDL Number:MFCD00007531