4-Bromo-2-ethyl-1-iodobenzene ≥97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:175278-30-5
:
Formula:C₈H₈BrI
MDL Number:MFCD00221458
:310.97 g/mol
Boiling Pt:84…86 °C (0.4 mmHg)
:Stabilised with copper
:GHS07
Storage Temperature:Ambient