Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl 4-bromobutyrate
tert-Butyl-4-bromobutyrate
tert-Butyl-4-bromobutyrate
tert-Butyl-4-bromobutyrate
Supplier:
COMBI-BLOCKS
Certificates
Expand image
tert-Butyl-4-bromobutyrate
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
110611-91-1
:
MFCD02179416
Formula:
C₇₁H₁₁₀NO₁₅P
:
1248.60 g/mol