Auftragserfassung
Germany
AuftragsverfolgungKontakt
Diethylethyl(phenyl)malonat

Diethylethyl(phenyl)malonat

Marketsource
Supplier:   Thermo Scientific
Diethylethyl(phenyl)malonat
Supplier:  Thermo Scientific
Marketsource
Certificates
Diethylethyl(phenyl)malonat

3 Options Available Below

Product Details & Documents
Synonyms:  Diethyl 2-ethyl-2-phenylmalonate
  • CAS Number:76-67-5
  • Formula:C₁₅H₂₀O₄
  • MDL Number:MFCD00040759
  • Molecular Weight:264.32 g/mol
  • EINECS:200-978-0
Product Family Options

Product Information

  • Packung
    VE
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.