Order Entry
Germany
ContactUsLinkComponent
Sodium deuteroxide 30% (w/w) in D₂O for NMR spectroscopy (D₂O 99+ atom%D)
Corrosive
Sodium deuteroxide 30% (w/w) in D₂O for NMR spectroscopy (D₂O 99+ atom%D)
Catalog # ACRO172890200
CAS Number:  
undefined
Corrosive
Sodium deuteroxide 30% (w/w) in D₂O for NMR spectroscopy (D₂O 99+ atom%D)
Catalog # ACRO172890200
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    20 g
  • Seq:
    3290

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • D-Enrichment:
    ≥99.0 %
  • Concentration:
    29 to 32 % w/w
  • Specific optical rotation:
    -2° to -8° (20°C, 589 nm) c=0.5 in Methanol

Specification Test Results

About this item

 :
 :41.01 g/mol
Density:1.46 g/cm³
Storage Temperature:Ambient
EINECS:237-825-2
UN:1824
ADR:8,II
 :GHS05
CAS Number:14014-06-3
Formula:NaOD
MDL Number:MFCD00037669