Order Entry
Germany
ContactUsLinkComponent
2-Isopropyl-3-methoxypyrazine
2-Isopropyl-3-methoxypyrazine
Catalog # EHERC14463700
Supplier:  EHRENSTORFER
CAS Number:  
2-Isopropyl-3-methoxypyrazine
Catalog # EHERC14463700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :152.1900 g/mol
EINECS:247-256-1
CAS Number:25773-40-4
Formula:C₈H₁₂N₂O
MDL Number:MFCD00006135