2-sec-Butyl-3-methoxypyrazine, Sigma-Aldrich®
Catalog # SIAL243116-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.2203 g/mol
:M-Clarity™ quality level = MQ100
EINECS:246-050-9
CAS Number:24168-70-5
Formula:C₉H₁₄N₂O
MDL Number:MFCD00006136