
Hafnium carbide ≥99.5% (metals basis excluding Zr) (max. 1% Zr)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12069-85-1
:
Formula:CHf
MDL Number:MFCD00016126
:190.54 g/mol
Melting Pt:3890 °C
Density:12.2
Storage Temperature:Ambient
EINECS:235-114-1
UN:3178
ADR:4.1,III
:GHS02